Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297669.1 | NP_001284598.1 | 5, 6 | 1021 | ||
NM_001297670.1 | NP_001284599.1 | 4, 5 | 911 | |||
NM_001297671.1 | NP_001284600.1 | 4, 5 | 647 | |||
NM_001297672.1 | NP_001284601.1 | 4, 5 | 647 | |||
NM_015149.4 | NP_055964.3 | 5, 6 | 1022 | |||
XM_011509339.2 | XP_011507641.1 | 6, 7 | 1068 | |||
XM_011509341.1 | XP_011507643.1 | 5, 6 | 975 | |||
XM_011509342.2 | XP_011507644.1 | 5, 6 | 975 | |||
XM_017000756.1 | XP_016856245.1 | 4, 5 | 496 | |||
GenBank mRNA | AB023176.1 | - | 4, 5 | 587 | ||
AF186779.1 | - | 4, 5 | 608 | |||
AF186780.1 | - | 5, 6 | 984 | |||
AK295179.1 | - | 4, 5 | 785 | |||
AK304293.1 | - | 4, 5 | 572 | |||
AK311701.1 | - | 4, 5 | 493 | |||
AK311716.1 | - | 4, 5 | 647 | |||
AK316526.1 | - | 5, 6 | 959 | |||
AL080117.1 | - | 3, 4 | 390 | |||
BC136591.1 | - | 5, 6 | 726 | 9020581 | ||
BX537416.1 | - | 5, 6 | 962 |