Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297673.1 | NP_001284602.1 | 4 | 1066 | ||
NM_001297674.1 | NP_001284603.1 | 4 | 1066 | |||
NM_015203.4 | NP_056018.2 | 5 | 1144 | |||
XM_005245033.2 | XP_005245090.1 | 5 | 1107 | |||
XM_006711240.2 | XP_006711303.1 | 5 | 1107 | |||
XM_011509344.2 | XP_011507646.1 | 5 | 1107 | |||
XM_017000763.1 | XP_016856252.1 | 4 | 1029 | |||
XM_017000764.1 | XP_016856253.1 | 5 | 482 | |||
XM_017000765.1 | XP_016856254.1 | 5 | 1107 | |||
XM_017000766.1 | XP_016856255.1 | 4 | 741 | |||
XM_017000767.1 | XP_016856256.1 | 4 | 1029 | |||
GenBank mRNA | AB007929.2 | - | 5 | 513 | ||
AK056707.1 | - | 5 | 551 | |||
AK125864.1 | - | 5 | 714 | |||
AK291703.1 | - | 5 | 562 | |||
AK304380.1 | - | 4 | 484 | |||
AK308148.1 | - | 4 | 475 | |||
BC045623.1 | - | 4 | 467 | 5269891 | ||
BX641025.1 | - | 6 | 1039 |