Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCAPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015261.2 | NP_056076.1 | 2 | 736 | ||
XM_011542719.2 | XP_011541021.1 | 2 | 160 | |||
GenBank mRNA | AK124878.1 | - | 2 | 736 | ||
BC098398.1 | - | 2 | 195 | 30528692 | ||
BC144524.1 | - | 2 | 420 | |||
D29954.1 | - | 2 | 158 |