Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015268.3 | NP_056083.3 | 19 | 2070 | ||
XM_005247245.4 | XP_005247302.1 | 19 | 2252 | |||
XM_017006036.1 | XP_016861525.1 | 6 | 527 | |||
GenBank mRNA | AB014578.2 | - | 19 | 2119 | ||
AK127112.1 | - | 3 | 158 | |||
AY779857.1 | - | 19 | 2070 | |||
BC043583.1 | - | 19 | 2070 | |||
BC151246.1 | - | 20 | 2140 | 9007477 |