Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED13L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015335.4 | NP_056150.1 | 2 | 342 | ||
XM_011538080.2 | XP_011536382.1 | 2 | 375 | |||
XM_011538081.2 | XP_011536383.1 | 2 | 375 | |||
XM_011538082.2 | XP_011536384.1 | 2 | 545 | |||
XM_017019090.1 | XP_016874579.1 | 2 | 375 | |||
GenBank mRNA | AF515599.1 | - | 2 | 342 | ||
AY338463.1 | - | 2 | 494 | |||
BC130422.1 | - | 2 | 338 | 40146453 |