Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WHRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173425.1 | NP_001166896.1 | 1 | 972 | ||
NM_015404.3 | NP_056219.3 | 1 | 972 | |||
XM_011518484.1 | XP_011516786.1 | 1 | 1025 | |||
XM_011518485.1 | XP_011516787.1 | 2 | 873 | |||
XM_011518486.1 | XP_011516788.1 | 1 | 1025 | |||
XM_011518487.1 | XP_011516789.1 | 1 | 1025 | |||
XM_011518488.1 | XP_011516790.1 | 1 | 1025 | |||
XM_011518489.2 | XP_011516791.1 | 1 | 1025 | |||
XM_011518491.2 | XP_011516793.1 | 1 | 1025 | |||
XM_011518492.1 | XP_011516794.1 | 1 | 1025 | |||
XM_011518494.2 | XP_011516796.1 | 1 | 1025 | |||
GenBank mRNA | AB040959.1 | - | 1 | 486 | ||
AK056190.1 | - | 1 | 966 | |||
BC014524.1 | - | 1472 | ||||
BC136416.1 | - | 1 | 721 | 9020404 | ||
BC142614.1 | - | 1 | 972 | 40147287 | ||
BC142684.1 | - | 1 | 947 | 40148465 |