Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KBTBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015483.2 | NP_056298.2 | 2, 3 | 837 | ||
XM_005249696.2 | XP_005249753.1 | 2, 3 | 693 | |||
XM_011515256.2 | XP_011513558.1 | 2, 3 | 572 | |||
GenBank mRNA | AF151831.1 | - | 1, 2 | 246 | ||
AK292802.1 | - | 2, 3 | 559 | |||
BC037887.2 | - | 2, 3 | 741 | 5274706 | ||
BC047107.1 | - | 2, 3 | 574 | 5275217 |