Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004539.3 | NP_004530.1 | 5, 6 | 652 | ||
XM_005266700.1 | XP_005266757.1 | 5, 6 | 430 | |||
GenBank mRNA | AJ000334.1 | - | 5, 6 | 497 | ||
AK129897.1 | - | 5, 6 | 455 | |||
AK222835.1 | - | 5, 6 | 499 | |||
AK293985.1 | - | 4, 5 | 432 | |||
AK294763.1 | - | 5, 6 | 483 | |||
BC001687.1 | - | 5, 6 | 501 | 2987840 | ||
D84273.1 | - | 5, 6 | 456 |