Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SENP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100409.2 | NP_001093879.1 | 16 | 3118 | ||
NM_015571.3 | NP_056386.2 | 17 | 3139 | |||
XM_006715457.3 | XP_006715520.1 | 18 | 2893 | |||
XM_011535746.2 | XP_011534048.1 | 16 | 2691 | |||
XM_011535747.2 | XP_011534049.1 | 16 | 3042 | |||
XM_017010719.1 | XP_016866208.1 | 17 | 2781 | |||
XM_017010720.1 | XP_016866209.1 | 17 | 2097 | |||
XM_017010721.1 | XP_016866210.1 | 17 | 2105 | |||
XM_017010722.1 | XP_016866211.1 | 18 | 2912 | |||
XM_017010723.1 | XP_016866212.1 | 18 | 2889 | |||
XM_017010724.1 | XP_016866213.1 | 18 | 3264 | |||
XM_017010725.1 | XP_016866214.1 | 15 | 3024 | |||
GenBank mRNA | AB018340.2 | - | 17 | 2167 | ||
AF196304.1 | - | 17 | 2123 | |||
AF306508.1 | - | 17 | 2138 | |||
AF307849.1 | - | 17 | 2198 | |||
AK021417.1 | - | 8 | 970 | |||
AK021949.1 | - | 1 | 62 | |||
AK126921.1 | - | 16 | 3118 | |||
BC028583.1 | - | 16 | 2325 | 4838569 |