Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSL1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015659.2 | NP_056474.2 | 3 | 381 | ||
GenBank mRNA | AJ007398.1 | - | 3 | 313 | ||
AK000822.1 | - | 3 | 325 | |||
AK295935.1 | - | 2 | 236 | |||
AL049999.1 | - | 3 | 368 | |||
AY154473.1 | - | 3 | 342 | |||
AY598331.1 | - | 3 | 308 | |||
BC016362.1 | - | 3 | 344 | |||
BC017899.1 | - | 3 | 330 | |||
BC019069.2 | - | 3 | 314 | |||
BC070325.1 | - | 3 | 332 | |||
BC107782.1 | - | 3 | 334 | |||
BC112228.1 | - | 3 | 346 | 8327696 | ||
BC113699.1 | - | 3 | 334 | 8322751 | ||
BI856275.1 | - | 3 | 381 |