Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT172 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015662.2 | NP_056477.1 | 5, 6 | 568 | ||
XM_005264254.1 | XP_005264311.1 | 5, 6 | 465 | |||
XM_006711986.3 | XP_006712049.1 | 6, 7 | 660 | |||
XM_006711987.1 | XP_006712050.1 | 5, 6 | 465 | |||
XM_011532757.2 | XP_011531059.1 | 5, 6 | 774 | |||
XM_011532758.1 | XP_011531060.1 | 5, 6 | 465 | |||
XM_017003790.1 | XP_016859279.1 | 6, 7 | 660 | |||
XM_017003791.1 | XP_016859280.1 | 4, 5 | 535 | |||
XM_017003792.1 | XP_016859281.1 | 5, 6 | 465 | |||
XM_017003793.1 | XP_016859282.1 | 4, 5 | 388 | |||
XM_017003794.1 | XP_016859283.1 | 2, 3 | 91 | |||
XM_017003795.1 | XP_016859284.1 | 5, 6 | 772 | |||
GenBank mRNA | AK055826.1 | - | 5, 6 | 461 | ||
AK299576.1 | - | 6, 7 | 630 | |||
AK310417.1 | - | 5, 6 | 490 | |||
AL110218.1 | - | 2, 3 | 124 | |||
BC137126.1 | - | 5, 6 | 458 | 9021123 | ||
BC142675.1 | - | 5, 6 | 530 | |||
BC142729.1 | - | 5, 6 | 530 |