Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198798.2 | NP_001185727.1 | 9 | 1213 | ||
NM_001198799.2 | NP_001185728.1 | 10 | 1258 | |||
NM_001198800.2 | NP_001185729.1 | 9 | 1174 | |||
NR_045564.1 | 9 | 1253 | ||||
XM_006717873.3 | XP_006717936.1 | 9 | 1415 | |||
XM_011539841.2 | XP_011538143.1 | 9 | 1038 | |||
XM_011539842.2 | XP_011538144.1 | 9 | 1415 | |||
XM_017016291.1 | XP_016871780.1 | 9 | 1038 | |||
XM_017016292.1 | XP_016871781.1 | 9 | 1415 | |||
XM_017016293.1 | XP_016871782.1 | 9 | 1038 | |||
XM_017016294.1 | XP_016871783.1 | 9 | 1415 | |||
XM_017016295.1 | XP_016871784.1 | 8 | 918 | |||
XM_017016296.1 | XP_016871785.1 | 7 | 819 | |||
XM_017016297.1 | XP_016871786.1 | 8 | 918 | |||
XM_017016298.1 | XP_016871787.1 | 8 | 1295 | |||
XM_017016299.1 | XP_016871788.1 | 8 | 918 | |||
XM_017016300.1 | XP_016871789.1 | 8 | 1295 | |||
GenBank mRNA | AF132952.1 | - | 10 | 1323 | ||
AK023436.1 | - | 9 | 1021 | |||
AK094170.1 | - | 10 | 1092 | |||
AK096409.1 | - | 7 | 1724 | |||
AK124364.1 | - | 2 | 1442 | |||
AK298302.1 | - | 9 | 1253 | |||
AL359339.1 | - | 4 | 332 | |||
AY013290.1 | - | 10 | 1323 | |||
BC012291.1 | - | 9 | 1060 | 3858865 |