Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016406.3 | NP_057490.2 | 6 | 794 | ||
XM_005245254.1 | XP_005245311.1 | 5 | 655 | |||
XM_017001450.1 | XP_016856939.1 | 7 | 639 | |||
XM_017001451.1 | XP_016856940.1 | 7 | 903 | |||
GenBank mRNA | AF151884.1 | - | 6 | 761 | ||
AF161504.1 | - | 6 | 780 | |||
AK292776.1 | - | 6 | 746 | |||
BC005187.1 | - | 6 | 786 | 3961569 |