Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCKIPSD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016453.3 | NP_057537.1 | 3 | 487 | ||
NM_184231.2 | NP_909119.1 | 3 | 487 | |||
XM_017006594.1 | XP_016862083.1 | 3 | 481 | |||
XM_017006595.1 | XP_016862084.1 | 3 | 495 | |||
XM_017006596.1 | XP_016862085.1 | 3 | 501 | |||
XM_017006597.1 | XP_016862086.1 | 3 | 245 | |||
XM_017006598.1 | XP_016862087.1 | 3 | 492 | |||
XM_017006599.1 | XP_016862088.1 | 3 | 507 | |||
XM_017006600.1 | XP_016862089.1 | 3 | 491 | |||
GenBank mRNA | AF178432.1 | - | 3 | 455 | ||
AF303581.1 | - | 3 | 426 | |||
AJ242655.2 | - | 3 | 455 | |||
AY453794.1 | - | 3 | 455 | |||
BC016052.1 | - | 3 | 371 | 4862348 | ||
BC026280.1 | - | 3 | 438 | 4730420 | ||
HY010884.1 | - | 3 | 488 |