Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYDIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198542.1 | NP_001185471.1 | 4 | 421 | ||
NM_001198543.1 | NP_001185472.1 | 4 | 391 | |||
NM_001270974.2 | NP_001257903.1 | 4 | 489 | |||
NM_017558.4 | NP_060028.2 | 4 | 489 | |||
XM_006721206.2 | XP_006721269.1 | 4 | 383 | |||
XM_011523146.2 | XP_011521448.1 | 4 | 416 | |||
XM_011523147.1 | XP_011521449.1 | 4 | 383 | |||
XM_011523148.1 | XP_011521450.1 | 4 | 456 | |||
XM_011523151.1 | XP_011521453.1 | 4 | 415 | |||
XM_017023346.1 | XP_016878835.1 | 4 | 661 | |||
GenBank mRNA | AK022933.1 | - | 4 | 423 | ||
AK299016.1 | - | 4 | 378 | |||
AK299348.1 | - | 4 | 421 | |||
AK308296.1 | - | 3 | 208 | |||
AL705531.1 | - | 4 | 433 | |||
BC028351.1 | - | 4 | 370 | |||
BP228881.1 | - | 4 | 408 | |||
DA758908.1 | - | 4 | 456 | |||
DB079205.1 | - | 4 | 456 |