Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017753.2 | NP_060223.2 | 5 | 885 | ||
NM_207299.1 | NP_997182.1 | 5 | 1018 | |||
GenBank mRNA | AK000307.1 | - | 5 | 885 | ||
AK225306.1 | - | 5 | 885 | |||
AK303718.1 | - | 3 | 765 | |||
AK303807.1 | - | 4 | 881 | |||
AK309115.1 | - | 3 | 594 | |||
AY304515.1 | - | 5 | 751 | |||
AY337718.1 | - | 4 | 631 | |||
BC022465.1 | - | 5 | 751 | 4797229 |