Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017805.2 | NP_060275.2 | 10 | 2609 | ||
XM_011527053.2 | XP_011525355.2 | 10 | 2729 | |||
XM_011527054.2 | XP_011525356.2 | 10 | 2729 | |||
XM_011527055.2 | XP_011525357.2 | 7 | 2435 | |||
XM_017026914.1 | XP_016882403.1 | 10 | 2729 | |||
XM_017026915.1 | XP_016882404.1 | 7 | 2435 | |||
GenBank mRNA | AK000408.1 | - | 6 | 724 | ||
AK123179.1 | - | 2 | 906 | |||
AY378097.1 | - | 10 | 2474 | |||
BC021860.1 | - | 6 | 739 | |||
BC028614.1 | - | 10 | 2481 | 4815880 | ||
BX389914.2 | - | 2 | 108 |