Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THG1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317824.1 | NP_001304753.1 | 3, 4 | 497 | ||
NM_001317825.1 | NP_001304754.1 | 3, 4 | 588 | |||
NM_001317826.1 | NP_001304755.1 | 4, 5 | 560 | |||
NM_017872.4 | NP_060342.2 | 3, 4 | 592 | |||
XM_017009603.1 | XP_016865092.1 | 3, 4 | 527 | |||
GenBank mRNA | AK000553.1 | - | 3, 4 | 568 | ||
AK021663.1 | - | 3, 4 | 543 | |||
AK023355.1 | - | 2, 3 | 206 | |||
AK223119.1 | - | 3, 4 | 561 | |||
AK304590.1 | - | 3, 4 | 468 | |||
AL136669.1 | - | 3, 4 | 560 | |||
AY463216.1 | - | 3, 4 | 666 | |||
BC001523.2 | - | 3, 4 | 538 | 3010614 | ||
BC001852.2 | - | 3, 4 | 538 | 3010614 | ||
BC023521.2 | - | 3, 4 | 558 | 4126840 | ||
CR533503.1 | - | 3, 4 | 540 |