Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTPAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018109.3 | NP_060579.3 | 9 | 2406 | ||
GenBank mRNA | AK022188.1 | - | 9 | 2386 |