Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288983.1 | NP_001275912.1 | 8 | 745 | ||
NM_001288984.1 | NP_001275913.1 | 9 | 931 | |||
NM_018115.3 | NP_060585.2 | 9 | 856 | |||
XM_005263102.4 | XP_005263159.1 | 9 | 912 | |||
XM_005263105.4 | XP_005263162.1 | 9 | 970 | |||
XM_017008354.1 | XP_016863843.1 | 8 | 774 | |||
XM_017008355.1 | XP_016863844.1 | 7 | 662 | |||
GenBank mRNA | AK001360.1 | - | 8 | 591 | ||
AK022683.1 | - | 7 | 510 | |||
AK300074.1 | - | 8 | 721 | |||
AK307540.1 | - | 9 | 839 | |||
BC048351.1 | - | 8 | 588 | |||
BC054040.1 | - | 9 | 797 | |||
BC063797.1 | - | 9 | 827 | |||
BC107895.1 | - | 9 | 836 | |||
CR749574.1 | - | 9 | 907 | |||
DN602051.1 | - | 3 | 153 |