Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113378.1 | NP_001106849.1 | 38 | 4527 | ||
NM_018193.2 | NP_060663.2 | 37 | 4347 | |||
XM_011521756.2 | XP_011520058.1 | 38 | 4551 | |||
XM_011521757.2 | XP_011520059.1 | 38 | 4547 | |||
XM_011521764.2 | XP_011520066.1 | 37 | 4371 | |||
XM_011521767.2 | XP_011520069.1 | 38 | 4521 | |||
GenBank mRNA | AB058697.1 | - | 24 | 4452 | ||
AK027564.1 | - | 26 | 3235 |