Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146688.1 | NP_001140160.1 | 6 | 669 | ||
NM_018433.5 | NP_060903.2 | 6 | 938 | |||
XM_006712051.3 | XP_006712114.1 | 5 | 641 | |||
XM_017004493.1 | XP_016859982.1 | 6 | 957 | |||
XM_017004494.1 | XP_016859983.1 | 6 | 727 | |||
GenBank mRNA | AB018285.2 | - | 6 | 701 | ||
AK304276.1 | - | 4 | 455 | |||
AL832150.1 | - | 6 | 704 | |||
BC038297.2 | - | 6 | 715 | |||
BX640698.1 | - | 6 | 739 | |||
BX647271.1 | - | 6 | 707 | |||
BX647752.1 | - | 6 | 767 | |||
BX648841.1 | - | 6 | 670 | |||
BX649109.1 | - | 6 | 708 | |||
CR749581.1 | - | 6 | 706 |