Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018462.4 | NP_060932.2 | 2 | 236 | ||
GenBank mRNA | AF161418.1 | - | 2 | 197 | ||
AK312155.1 | - | 2 | 210 | |||
AY148219.1 | - | 2 | 236 | |||
AY148220.1 | - | 2 | 236 | |||
BC001067.1 | - | 2 | 212 | 2823162 | ||
BC007929.1 | - | 2 | 205 | 3688264 | ||
BC019303.1 | - | 2 | 203 | 2823162 | ||
BC061515.1 | - | 2 | 210 |