Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOSPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019556.2 | NP_062456.1 | 5 | 741 | ||
XM_011531366.2 | XP_011529668.1 | 5 | 695 | |||
XM_017029653.1 | XP_016885142.1 | 6 | 934 | |||
GenBank mRNA | AK307196.1 | - | 4 | 568 | ||
AK316570.1 | - | 5 | 616 | |||
AL137163.1 | - | 5 | 664 | |||
BC005700.1 | - | 5 | 644 | 3450155 |