Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBMX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164803.1 | NP_001158275.1 | 8 | 961 | ||
GenBank mRNA | AK097941.1 | - | 1 | 662 | ||
AK304868.1 | - | 8 | 906 | |||
BX647131.1 | - | 8 | 3794 |
Gene Symbol: RBMXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162536.2 | NP_001156008.1 | 3 | 3010 | ||
NM_019610.5 | NP_062556.2 | 2 | 2910 | |||
GenBank mRNA | AL832554.1 | - | 2 | 2763 | ||
AL832785.1 | - | 1 | 1854 | |||
BC012942.1 | - | 2 | 2569 | 3875565 |