Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193557.1 | NP_001180486.1 | 5 | 373 | ||
NM_020189.5 | NP_064574.1 | 5 | 445 | |||
NR_036471.1 | 5 | 443 | ||||
NR_036472.1 | 5 | 490 | ||||
GenBank mRNA | AF173296.1 | - | 5 | 515 | ||
AF201940.1 | - | 5 | 460 | |||
AK000487.1 | - | 5 | 349 | |||
AK095651.1 | - | 5 | 489 | |||
AK290752.1 | - | 5 | 389 | |||
BC007870.2 | - | 5 | 361 | 4303290 | ||
BX537648.1 | - | 2 | 269 | |||
BX648621.1 | - | 3 | 1194 | |||
DA548665.1 | - | 5 | 319 | |||
DA762166.1 | - | 5 | 425 | |||
DB168034.1 | - | 5 | 445 |