Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020232.4 | NP_064617.2 | 5 | 625 | ||
NM_147163.1 | NP_671692.1 | 5 | 483 | |||
GenBank mRNA | AF068295.1 | - | 5 | 483 | ||
AF161378.1 | - | 5 | 482 | |||
AF168712.1 | - | 5 | 557 | |||
AF276707.1 | - | 5 | 1185 | |||
AK057005.1 | - | 5 | 557 | |||
AK225882.1 | - | 5 | 592 | |||
BC013356.2 | - | 5 | 548 | 3640209 | ||
CR457181.1 | - | 4 | 385 |