Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020697.3 | NP_065748.1 | 2 | 789 | ||
GenBank mRNA | AB032970.1 | - | 2 | 594 | ||
AK293096.1 | - | 1 | 733 | |||
BC027932.1 | - | 2 | 762 | 5199736 | ||
BC034778.1 | - | 2 | 789 | 5175712 |