Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020718.3 | NP_065769.3 | 12 | 1886 | ||
XM_017023497.1 | XP_016878986.1 | 12 | 3412 | |||
XM_017023498.1 | XP_016878987.1 | 11 | 1199 | |||
XM_017023499.1 | XP_016878988.1 | 9 | 1003 | |||
XM_017023500.1 | XP_016878989.1 | 12 | 1917 | |||
GenBank mRNA | AB033029.1 | - | 11 | |||
AJ586135.2 | - | 12 | 1886 | |||
AK126752.1 | - | 12 | 1570 | |||
BX648357.1 | - | 3 | 274 |