Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020718.3 | NP_065769.3 | 9, 10 | 1616 | ||
XM_017023496.1 | XP_016878985.1 | 9, 10 | 1647 | |||
XM_017023497.1 | XP_016878986.1 | 9, 10 | 3142 | |||
XM_017023498.1 | XP_016878987.1 | 8, 9 | 929 | |||
XM_017023499.1 | XP_016878988.1 | 6, 7 | 733 | |||
XM_017023500.1 | XP_016878989.1 | 9, 10 | 1647 | |||
GenBank mRNA | AJ586135.2 | - | 9, 10 | 1616 | ||
BM842261.1 | - | 3, 4 | 313 |