Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRGAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020762.2 | NP_065813.1 | 2 | 175 | ||
XM_005269042.3 | XP_005269099.1 | 2 | 564 | |||
XM_011538580.1 | XP_011536882.1 | 3 | 283 | |||
XM_011538581.2 | XP_011536883.1 | 3 | 1788 | |||
GenBank mRNA | AB037725.1 | - | 1 | 1466 | ||
BC029919.1 | - | 2 | 133 | |||
BC053903.1 | - | 2 | 175 | 6526787 |