Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006657.1 | NP_001006658.1 | 22, 23 | 2681 | ||
NM_020779.3 | NP_065830.2 | 21, 22 | 2648 | |||
XM_011533007.2 | XP_011531309.1 | 11, 12 | 1373 | |||
GenBank mRNA | AB037757.2 | - | 22, 23 | 2689 | ||
AK122917.1 | - | 21, 22 | 2573 | |||
BC036659.2 | - | 21, 22 | 2647 | 5266940 |