Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIPA1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020808.4 | NP_065859.3 | 1 | 1159 | ||
XM_005273211.2 | XP_005273268.1 | 3 | 1496 | |||
XM_005273212.4 | XP_005273269.1 | 3 | 1303 | |||
XM_005273213.4 | XP_005273270.1 | 2 | 1196 | |||
XM_011544243.2 | XP_011542545.1 | 4 | 1352 | |||
XM_017001896.1 | XP_016857385.1 | 3 | 1249 | |||
XM_017001897.1 | XP_016857386.1 | 3 | 1496 | |||
XM_017001898.1 | XP_016857387.1 | 3 | 1270 | |||
GenBank mRNA | AB037810.1 | - | 1 | 363 | ||
AY168879.1 | - | 1 | 1119 | |||
BC006013.1 | - | 1 | 1033 | |||
BC150334.1 | - | 1 | 374 | 8860510 |