Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029939.3 | NP_001025110.2 | 3 | 340 | ||
NM_001167583.2 | NP_001161055.1 | 2 | 181 | |||
NM_001174157.1 | NP_001167628.1 | 2 | 258 | |||
NM_001174158.1 | NP_001167629.1 | 2 | 181 | |||
NM_001289394.1 | NP_001276323.1 | 3 | 417 | |||
NM_020863.3 | NP_065914.2 | 2 | 258 | |||
NR_110323.1 | 2 | 258 | ||||
XM_011517203.1 | XP_011515505.1 | 2 | 155 | |||
XM_011517206.1 | XP_011515508.1 | 2 | 179 | |||
XM_017013716.1 | XP_016869205.1 | 1 | 53 | |||
GenBank mRNA | AB167738.1 | - | 2 | 247 | ||
AB167739.1 | - | 3 | 406 | |||
AB167740.1 | - | 3 | 333 | |||
AB167741.1 | - | 2 | 174 | |||
AK094089.1 | - | 2 | 241 | |||
AK300133.1 | - | 2 | 170 | |||
AK300324.1 | - | 2 | 177 | |||
AK300471.1 | - | 2 | 243 | |||
AK311494.1 | - | 2 | 179 | |||
AL832066.1 | - | 2 | 258 | |||
BC012989.2 | - | 2 | 172 | |||
BC098405.1 | - | 2 | 181 | |||
BC101766.1 | - | 2 | 171 | 8069272 | ||
BC101768.1 | - | 2 | 171 | 8069274 | ||
BC143519.1 | - | 2 | 172 | 9052028 |