Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029939.3 | NP_001025110.2 | 5 | 868 | ||
NM_001167583.2 | NP_001161055.1 | 4 | 709 | |||
NM_001174158.1 | NP_001167629.1 | 4 | 709 | |||
NM_001289394.1 | NP_001276323.1 | 5 | 945 | |||
NM_020863.3 | NP_065914.2 | 4 | 786 | |||
NR_110323.1 | 4 | 786 | ||||
XM_011517203.1 | XP_011515505.1 | 4 | 683 | |||
XM_011517204.2 | XP_011515506.1 | 3 | 506 | |||
XM_011517206.1 | XP_011515508.1 | 4 | 707 | |||
XM_017013716.1 | XP_016869205.1 | 3 | 581 | |||
GenBank mRNA | AB040918.1 | - | 2 | 184 | ||
AB167738.1 | - | 4 | 775 | |||
AB167739.1 | - | 5 | 934 | |||
AB167740.1 | - | 5 | 861 | |||
AB167741.1 | - | 4 | 702 | |||
AK300133.1 | - | 4 | 698 | |||
AK300324.1 | - | 4 | 705 | |||
AK300471.1 | - | 4 | 771 | |||
AK311494.1 | - | 4 | 707 | |||
AL832066.1 | - | 4 | 786 | |||
BC012989.2 | - | 4 | 700 | |||
BC025423.2 | - | 2 | 378 | |||
BC098405.1 | - | 4 | 709 | |||
BC101766.1 | - | 4 | 699 | 8069272 | ||
BC101768.1 | - | 4 | 699 | 8069274 |