Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304363.1 | NP_001291292.1 | 2 | 742 | ||
NM_001304364.1 | NP_001291293.1 | 2 | 871 | |||
NM_020924.3 | NP_065975.1 | 2 | 784 | |||
GenBank mRNA | AB046792.1 | - | 1 | 1902 | ||
AF323460.2 | - | 1 | 669 | |||
AK057445.1 | - | 2 | 871 | |||
AK058083.1 | - | 1 | 311 | |||
BC018748.1 | - | 2 | 733 | 4553653 |