Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRINP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021165.3 | NP_066988.1 | 8 | 1989 | ||
XM_005245379.1 | XP_005245436.1 | 9 | 2569 | |||
XM_011509826.1 | XP_011508128.1 | 6 | 1816 | |||
GenBank mRNA | AB051534.1 | - | 8 | 1982 | ||
AB161694.1 | - | 8 | 1982 | |||
AK098386.1 | - | 1 | 196 | |||
AK123336.1 | - | 5 | 1424 | |||
AK296524.1 | - | 9 | 2065 | |||
AL035289.1 | - | 7 | 1563 | |||
BC028036.1 | - | 8 | 1881 | 5242899 |