Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AVPI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021732.2 | NP_068378.2 | 3 | 874 | ||
XM_017016494.1 | XP_016871983.1 | 3 | 1034 | |||
GenBank mRNA | AF131791.1 | - | 3 | 874 | ||
AF218019.1 | - | 3 | 829 | |||
AF241786.1 | - | 3 | 876 | |||
AK024533.1 | - | 3 | 846 | |||
AK222493.1 | - | 3 | 847 | |||
AK223099.1 | - | 3 | 822 | |||
BC000877.1 | - | 3 | 485 | 3461724 |