Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RINT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021930.4 | NP_068749.3 | 6 | 938 | ||
XM_005250524.3 | XP_005250581.1 | 2 | 126 | |||
XM_017012496.1 | XP_016867985.1 | 6 | 717 | |||
XM_017012497.1 | XP_016867986.1 | 6 | 813 | |||
GenBank mRNA | AF317622.1 | - | 6 | 817 | ||
AK055508.1 | - | 6 | 853 | |||
AK057757.1 | - | 3 | 339 | |||
BC068483.1 | - | 6 | 939 | 5270834 |