Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198713.1 | NP_001185642.1 | 2 | 218 | ||
NM_001256782.1 | NP_001243711.1 | 3 | 315 | |||
NM_001256822.1 | NP_001243751.1 | 2 | 287 | |||
NM_004574.4 | NP_004565.1 | 2 | 418 | |||
NM_080415.3 | NP_536340.1 | 2 | 189 | |||
NM_080416.3 | NP_536341.1 | 2 | 189 | |||
NR_037155.2 | 2 | 418 | ||||
NR_104196.1 | 2 | 418 | ||||
NR_104197.1 | 2 | 189 | ||||
XM_006721949.2 | XP_006722012.1 | 3 | 247 | |||
GenBank mRNA | AB002110.1 | - | 2 | 221 | ||
AB008753.1 | - | 2 | 253 | |||
AF035811.1 | - | 2 | 254 | |||
AF073312.1 | - | 2 | 249 | |||
AF176379.1 | - | 2 | 155 | |||
AK091065.1 | - | 2 | 156 | |||
AK094579.1 | - | 2 | 940 | |||
AK126631.1 | - | 1 | 1656 | |||
AK294094.1 | - | 2 | 221 | |||
AK295527.1 | - | 2 | 142 | |||
AK301914.1 | - | 3 | 315 | |||
AK302146.1 | - | 2 | 218 | |||
AK314528.1 | - | 2 | 301 | |||
AK315045.1 | - | 2 | 265 | |||
AK315396.1 | - | 2 | 143 | |||
BC018055.1 | - | 2 | 106 | |||
BC018056.1 | - | 2 | 132 | 4811662 | ||
BC128515.1 | - | 2 | 251 | |||
CR457111.1 | - | 2 | 125 | |||
D89278.1 | - | 2 | 125 | |||
DA076433.1 | - | 2 | 368 | |||
DA180095.1 | - | 2 | 418 | |||
DA343015.1 | - | 2 | 287 | |||
U88829.1 | - | 2 | 250 | |||
U88870.1 | - | 2 | 135 |