Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130820.2 | NP_001124292.1 | 4 | 933 | ||
NM_001130821.2 | NP_001124293.1 | 5 | 1054 | |||
NM_001130822.2 | NP_001124294.1 | 4 | 946 | |||
NM_001287525.1 | NP_001274454.1 | 6 | 1096 | |||
NM_001287526.1 | NP_001274455.1 | 3 | 856 | |||
NM_022777.3 | NP_073614.1 | 5 | 1023 | |||
GenBank mRNA | AK023287.1 | - | 6 | 1054 | ||
AK024179.1 | - | 5 | 949 | |||
AK298888.1 | - | 4 | 887 | |||
AL157469.1 | - | 2 | 635 |