Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022830.2 | NP_073741.2 | 2 | 286 | ||
GenBank mRNA | AB101208.1 | - | 2 | 200 | ||
AK026000.1 | - | 2 | 193 | |||
AK292610.1 | - | 2 | 285 | |||
BC110910.1 | - | 2 | 175 | 6141937 | ||
BC128263.1 | - | 2 | 179 | 40118672 |