Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAPLN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023002.2 | NP_075378.1 | 5 | 2799 | ||
GenBank mRNA | AY262756.1 | - | 5 | 2763 | ||
BC142644.1 | - | 5 | 2799 | 40147937 | ||
BC142698.1 | - | 5 | 2808 | 40148775 |