Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024652.4 | NP_078928.3 | 20 | 3060 | ||
XM_005254979.3 | XP_005255036.1 | 6 | 718 | |||
XM_011522012.2 | XP_011520314.1 | 22 | 3334 | |||
XM_011522013.2 | XP_011520315.1 | 20 | 3043 | |||
XM_011522014.2 | XP_011520316.1 | 20 | 3258 | |||
XM_011522015.2 | XP_011520317.1 | 15 | 3853 | |||
XM_011522016.2 | XP_011520318.1 | 18 | 2535 | |||
XM_011522017.2 | XP_011520319.1 | 6 | 725 | |||
XM_011522018.2 | XP_011520320.1 | 6 | 720 | |||
XM_011522019.2 | XP_011520321.1 | 20 | 3258 | |||
XM_017022570.1 | XP_016878059.1 | 10 | 1307 | |||
GenBank mRNA | AB058693.1 | - | 6 | 765 | ||
BC144705.1 | - | 19 | 2612 | |||
BC146769.1 | - | 6 | 765 | |||
BC171802.1 | - | 19 | 2734 | |||
DQ013130.1 | - | 19 | 2698 |