Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024652.4 | NP_078928.3 | 21 | 3456 | ||
XM_005254979.3 | XP_005255036.1 | 7 | 1114 | |||
XM_011522012.2 | XP_011520314.1 | 23 | 3730 | |||
XM_011522013.2 | XP_011520315.1 | 21 | 3439 | |||
XM_011522014.2 | XP_011520316.1 | 21 | 3654 | |||
XM_011522015.2 | XP_011520317.1 | 16 | 4249 | |||
XM_011522016.2 | XP_011520318.1 | 19 | 2931 | |||
XM_011522017.2 | XP_011520319.1 | 7 | 1121 | |||
XM_011522018.2 | XP_011520320.1 | 7 | 1116 | |||
XM_011522019.2 | XP_011520321.1 | 21 | 3654 | |||
XM_017022570.1 | XP_016878059.1 | 11 | 1703 | |||
GenBank mRNA | AB058693.1 | - | 7 | 1161 | ||
AK131075.1 | - | 2 | 389 | |||
BC144705.1 | - | 20 | 3008 | |||
BC146769.1 | - | 7 | 1161 | |||
BC171802.1 | - | 20 | 3130 | |||
DQ013130.1 | - | 20 | 3094 |