Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGNBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024835.4 | NP_079111.1 | 14 | 2803 | ||
XM_005257689.2 | XP_005257746.1 | 14 | 2785 | |||
XM_011525265.2 | XP_011523567.1 | 13 | 2548 | |||
XM_011525266.1 | XP_011523568.1 | 13 | 2671 | |||
XM_017025107.1 | XP_016880596.1 | 13 | 2516 | |||
GenBank mRNA | AF126964.1 | - | 13 | 2534 | ||
AK026214.1 | - | 6 | 1437 | |||
AK290687.1 | - | 13 | 2559 |