Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193434.1 | NP_001180363.1 | 6 | 840 | ||
NM_001193435.1 | NP_001180364.1 | 4 | 644 | |||
NM_024889.4 | NP_079165.3 | 6 | 740 | |||
NM_182601.1 | NP_872407.1 | 5 | 957 | |||
XM_005270162.2 | XP_005270219.1 | 6 | 879 | |||
XM_005270163.2 | XP_005270220.1 | 6 | 879 | |||
XM_011540171.1 | XP_011538473.1 | 6 | 879 | |||
XM_011540172.1 | XP_011538474.1 | 7 | 647 | |||
XM_011540173.1 | XP_011538475.1 | 6 | 879 | |||
XM_011540174.2 | XP_011538476.1 | 6 | 879 | |||
XM_011540175.1 | XP_011538477.1 | 6 | 818 | |||
XM_011540176.1 | XP_011538478.1 | 4 | 416 | |||
XM_017016666.1 | XP_016872155.1 | 6 | 879 | |||
GenBank mRNA | AK126354.1 | - | 4 | 416 | ||
AK128811.1 | - | 6 | 740 | |||
AK292867.1 | - | 6 | 612 | |||
BC037870.1 | - | 5 | 962 | 5273287 | ||
FJ236306.1 | - | 5 | 962 |