Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35G2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097599.1 | NP_001091068.1 | 2 | 1321 | ||
NM_001097600.1 | NP_001091069.1 | 2 | 1172 | |||
NM_025246.2 | NP_079522.2 | 2 | 1586 | |||
XM_006713773.3 | XP_006713836.1 | 2 | 3738 | |||
XM_011513214.2 | XP_011511516.1 | 2 | 1925 | |||
XM_017007289.1 | XP_016862778.1 | 2 | 1049 | |||
XM_017007290.1 | XP_016862779.1 | 2 | 1248 | |||
XM_017007291.1 | XP_016862780.1 | 2 | 1054 | |||
GenBank mRNA | AK314079.1 | - | 2 | 1481 | ||
AL136661.1 | - | 2 | 1467 | |||
BC000111.1 | - | 2 | 1062 | 3508204 | ||
BC022557.1 | - | 2 | 1107 | 4793991 | ||
BC050423.1 | - | 2 | 1586 |