Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004596.4 | NP_004587.1 | 1, 2 | 632 | ||
GenBank mRNA | AK313182.1 | - | 1, 2 | 245 | ||
BC000405.2 | - | 1, 2 | 612 | 2822987 | ||
BC008290.1 | - | 1, 2 | 196 | 3677365 | ||
CN286572.1 | - | 1, 2 | 634 | |||
X06347.1 | - | 1, 2 | 202 |