Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032283.2 | NP_115659.1 | 8 | 2080 | ||
GenBank mRNA | AK056427.1 | - | 8 | 1832 | ||
AL512765.1 | - | 8 | 1685 | |||
BC066776.1 | - | 8 | 2080 | 30342061 |